  • Search Results
  • Overcharging in biological systems: reversal of electrophoretic mobility of aqueous polyaspartate by multivalent cations.
Overcharging in biological systems: reversal of electrophoretic mobility of aqueous polyaspartate by multivalent cations.
Physical review letters (2012-06-12)
Anna Kubíčková, Tomáš Křížek, Pavel Coufal, Mario Vazdar, Erik Wernersson, Jan Heyda, Pavel Jungwirth
ABSTRACT
Charge reversal as an extreme case of charge compensation is directly observed by capillary electrophoresis for a negatively charged peptide in aqueous solutions of trivalent cations. Atomistic and coarse-grained simulations provide molecular interpretation of this effect showing that it is largely of electrostatic origin with a minor contribution of chemical specificity of the salt ions.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.