  • Search Results
  • Sildenafil improves long-term effect of endothelial progenitor cell-based treatment for monocrotaline-induced rat pulmonary arterial hypertension.
Sildenafil improves long-term effect of endothelial progenitor cell-based treatment for monocrotaline-induced rat pulmonary arterial hypertension.
Cytotherapy (2013-01-17)
Chia-Hung Yen, Tzu-Hsien Tsai, Steve Leu, Yung-Lung Chen, Li-Teh Chang, Han-Tan Chai, Sheng-Ying Chung, Sarah Chua, Ching-Yen Tsai, Hsueh-Wen Chang, Sheung-Fat Ko, Cheuk-Kwan Sun, Hon-Kan Yip
ABSTRACT
We hypothesized that the long-term therapeutic effect of combined sildenafil and bone marrow-derived endothelial progenitor cells (BMDEPCs) on monocrotaline (MCT)-induced rat pulmonary arterial hypertension (PAH) is superior to either treatment alone. Male Sprague-Dawley rats (n = 40) were equally divided into normal controls, MCT (65 mg/kg, subcutaneously) only, MCT + sildenafil (25 mg/kg/day, orally), MCT + BMDEPCs (2.0 × 10(6) autologous cells, intravenously) and MCT + sildenafil+ BMDEPCs. BMDEPCs and sildenafil were given on day 21 after MCT administration. Animals were sacrificed by day 90 after MCT administration. The apoptotic (caspase 3, Bax) and inflammatory (tumor necrosis factor-α, matrix metalloproteinase-9) biomarkers in right ventricle and lung and pulmonary expressions of fibrotic biomarkers (transforming growth factor-β, p-Smad3) and connexin 43 protein were lower in monotherapy groups (i.e., MCT + sildenafil and MCT + BMDEPCs) and further decreased in normal controls and combined treatment groups (i.e., MCT + sildenafil + BMDEPCs) compared with untreated animals (i.e., MCT only) (all P < 0.01). Expressions of anti-fibrotic biomarkers (bone morphogenetic protein-2, p-Smad1/5) and numbers of alveolar sacs and arterioles in lung were higher in monotherapy groups and further increased in normal controls and combined treatment groups compared with untreated animals (all P < 0.005). In right ventricle, connexin 43 and α-myosin heavy chain (MHC) expressions were higher in the monotherapy groups and further elevated in normal controls and combined treatment groups compared with untreated animals, whereas β-MHC exhibited the opposite pattern (all P < 0.01). Right ventricular systolic pressure and weight were lower in the monotherapy animals and further reduced in normal controls and combined treatment groups compared with untreated animals (all P < 0.0001). Combined therapy with BMDEPCs and sildenafil was superior to either treatment alone in attenuating rodent MCT-induced PAH.
Sigma-Aldrich
Crotaline
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.