  • Search Results
  • Effect of thienorphine on the isolated uterine strips from pregnant rats.
Effect of thienorphine on the isolated uterine strips from pregnant rats.
European journal of pharmacology (2013-02-19)
Peilan Zhou, Lingdi Yan, Zheng Yong, Gang Yu, Huajin Dong, Hui Yan, Ruibin Su, Zehui Gong
ABSTRACT
Opioid dependence is a serious worldwide health problem. Buprenorphine was used as an alternative to methadone for the treatment of opioid dependence, especially for pregnant women. Thienorphine was a partial opioid agonist with long-lasting antinociceptive effect and high oral bioavailability compared with its analogue buprenorphine. Till now, there was still no research about the effect of thienorphine on the isolated uterine muscles. This study examined the effects of thienorphine on the isolated rat oestrus and pregnant uterine strips. Area under the curve (AUC), amplitude and frequency were studied. Thienorphine induced a concentration-dependent decrease in the frequency and amplitude of the contraction on the isolated oestrus and pregnant uterine strips. Thienorphine exhibited less inhibition on the contractile amplitude of the isolated uterine strips from pregnant rats with the IC50 of 54.11 ± 7.41 μΜ, compared with buprenorphine (IC50, 19.42 ± 2.34 μΜ). In addition, thienorphine also exhibited less inhibition on the contractile frequency of the isolated uterine strips from pregnant rats, with the IC50 of 70.68 ± 12.44 μΜ, compared with buprenorphine (IC50, 19.20 ± 3.87 μΜ). On the isolated uterine muscle from pregnant rats, the AUC was decreased by thienorphine but was less potent than buprenorphine, the IC50 was 37.31 ± 7.43 μΜ for thienorphine and 13.52 ± 2.03 μΜ for buprenorphine. Thienorphine exhibited longer duration on the isolated rat pregnant uterine strips than buprenorphine. Thienorphine has less influence and longer duration on the isolated rat uterine muscles during pregnancy, which may be a new useful candidate for the opioid dependent pregnant women.
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride dihydrate, for molecular biology, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis