  • Search Results
  • Synthesis of alkylated potato starch derivatives and their potential in the aqueous solubilization of benzo[a]pyrene.
Synthesis of alkylated potato starch derivatives and their potential in the aqueous solubilization of benzo[a]pyrene.
Carbohydrate polymers (2013-03-08)
Ana-Maria Rosu, Catherine Rafin, Gheorghe Surpateanu, Gheorghe Brabie, Doru Neculai Miron, Etienne Veignie
ABSTRACT
For the development of renewable bioproducts able to solubilize organic persistent pollutant such as benzo[a]pyrene (BaP), modified potato starch was synthesized by alkylation. The addition of alkyl chains was performed with three different alkylation agents: epoxyalkane, alkenyl succinic anhydride and 1,4-butane sultone. Twelve alkylated starches were obtained with different molar substitutions (MS) and various alkyl chain lengths (to three carbons up to sixteen). The chemical structural characteristics were investigated by methods of (1)H NMR and FTIR. In comparison with the native starch, the ether modified starches showed in general an enhancement of their aqueous solubility whereas the ester modified starches stimulated the BaP aqueous solubilization. Indeed, the compounds P6 and P12, which increased 40-fold the BaP aqueous concentration, present high surfactant properties.
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
Supelco
Benzo[a]pyrene solution, certified reference material, 100 μg/mL in methylene chloride
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Sigma-Aldrich
1,4-Butane sultone, ≥99%
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®
Supelco
Ethyl ether solution, certified reference material, 2000 μg/mL in methanol
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis