  • Search Results
  • Potentiation of GABAA receptor activity by volatile anaesthetics is reduced by α5GABAA receptor-preferring inverse agonists.
Potentiation of GABAA receptor activity by volatile anaesthetics is reduced by α5GABAA receptor-preferring inverse agonists.
British journal of anaesthesia (2013-03-29)
I Lecker, Y Yin, D S Wang, B A Orser
ABSTRACT
Animal studies have shown that memory deficits in the early post-anaesthetic period can be prevented by pre-treatment with an inverse agonist that preferentially inhibits α5 subunit-containing γ-aminobutyric acid type A (α5GABA(A)) receptors. The goal of this in vitro study was to determine whether inverse agonists that inhibit α5GABA(A) receptors reduce anaesthetic potentiation of GABAA receptor activity. Cultures of hippocampal neurones were prepared from Swiss white mice, wild-type mice (genetic background C57BL/6J and Sv129Ev) and α5GABA(A)receptor null mutant (Gabra5-/-) mice. Whole-cell voltage clamp techniques were used to study the effects of the α5GABA(A) receptor-preferring inverse agonists L-655,708 and MRK-016 on anaesthetic potentiation of GABA-evoked currents. L-655,708 (50 nM) reduced sevoflurane potentiation of GABA-evoked current in wild-type neurones but not Gabra5-/- neurones, and produced a rightward shift in the sevoflurane concentration-response plot [sevoflurane EC50: 1.9 (0.1) mM; sevoflurane+L-655,708 EC(50): 2.4 (0.2) mM, P<0.05]. Similarly, L-655,708 (50 nM) reduced isoflurane potentiation of GABA-evoked current [isoflurane: 4.0 (0.6) pA pF(-1); isoflurane+L-655,708: 3.1 (0.5) pA pF(-1), P<0.01]. MRK-016 also reduced sevoflurane and isoflurane enhancement of GABA-evoked current [sevoflurane: 1.5 (0.1) pA pF(-1); sevoflurane+MRK-016 (10 nM): 1.2 (0.1) pA pF(-1), P<0.05; isoflurane: 3.5 (0.3) pA pF(-1); isoflurane+MRK-016 (1 nM): 2.9 (0.2) pA pF(-1), P<0.05]. L-655,708 and MRK-016 reduced the potentiation by inhaled anaesthetics of GABAA receptor activated by a low concentration of GABA. Future studies are required to determine whether this effect contributes to the memory preserving properties of inverse agonists after anaesthesia.
Sigma-Aldrich
MRK-016, ≥98% (HPLC)
Sevoflurane, European Pharmacopoeia (EP) Reference Standard