  • Search Results
  • Gain-switched pulses from InGaAs ridge-quantum-well lasers limited by intrinsic dynamical gain suppression.
Gain-switched pulses from InGaAs ridge-quantum-well lasers limited by intrinsic dynamical gain suppression.
Optics express (2013-04-03)
Shaoqiang Chen, Masahiro Yoshita, Takashi Ito, Toshimitsu Mochizuki, Hidefumi Akiyama, Hiroyuki Yokoyama
ABSTRACT
Gain-switched pulses of InGaAs double-quantum-well lasers fabricated from identical epitaxial laser wafers were measured under both current injection and optical pumping conditions. The shortest output pulse widths were nearly identical (about 40 ps) both for current injection and optical pumping; this result attributed the dominant pulse-width limitation factor to the intrinsic gain properties of the lasers. We quantitatively compared the experimental results with theoretical calculations based on rate equations incorporating gain nonlinearities. Close consistency between the experimental data and the calculations was obtained only when we assumed a dynamically suppressed gain value deviated from the steady-state gain value supported by standard microscopic theories.
Sigma-Aldrich
Indium, wire, diam. 2.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 0.5 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, pieces, 99.99% trace metals basis
Sigma-Aldrich
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, powder, 99.99% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, bars, 99.95%
Sigma-Aldrich
Indium, powder, −60 mesh, 99.999% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gallium arsenide, (single crystal substrate), <100>, diam. × thickness 2 in. × 0.5 mm