MilliporeSigma
Frataxin: a protein in search for a function.
Journal of neurochemistry (2013-07-24)
Annalisa Pastore, Helene Puccio
ABSTRACT
Reduced levels of the protein frataxin cause the neurodegenerative disease Friedreich's ataxia. Pathology is associated with disruption of iron-sulfur cluster biosynthesis, mitochondrial iron overload, and oxidative stress. Frataxin is a highly conserved iron-binding protein present in most organisms. Despite the intense interest generated since the determination of its pathology, identification of the cellular function of frataxin has so far remained elusive. In this review, we revisit the most significant milestones that have led us to our current understanding of frataxin and its functions. The picture that emerges is that frataxin is a crucial element of one of the most essential cellular machines specialized in iron-sulfur cluster biogenesis. Future developments, therefore, can be expected from further advancements in our comprehension of this machine.
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis
Sigma-Aldrich
Sulfur, 99.998% trace metals basis
Sigma-Aldrich
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
Iron, ≥99%, reduced, powder (fine)
Sigma-Aldrich
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Iron, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Sigma-Aldrich
Iron, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
Iron, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Supelco
Sulfur, PESTANAL®, analytical standard
Sigma-Aldrich
Carbonyl iron, ≥97% Fe basis
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Iron, rod, diam. 6.3 mm, 99.98% trace metals basis