[Effect of glyphosate on the energy exchange in carp organs].
Ukrains'kyi biokhimichnyi zhurnal (1999 ) (2013-08-14)
A A Zhidenko, E V Bibchuk, E V Barbukho
ABSTRACT
The use of glyphosate as a herbicide in agriculture can lead to the presence of its residues and metabolites (aminomethylphosphonic acid) in food for human consumption and pose a threat to health. The effect of these herbicides on the fish organism at the biochemical level has been insufficiently studied. We studied changes in the content of adenine nucleotides, enzyme activity, quantitative indexes of energy metabolism substrates in carp under the action of glyphosate. It has been found that proteins are the major energy substrate under the influence of glyphosate in the liver, brain, white muscle of carp yearlings. Glyphosphate decreases energy metabolism in the brain of carp and increases it in the white muscles. The growth of activity of catabolic enzymes in the liver under the influence of glyphosate can be attributed to the adaptive remodelling of metabolic pathways for homeostasis and enantiostasis in response to herbicides.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Supelco
Glyphosate, PESTANAL®, analytical standard
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)
SAFC
Glycine
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Glycogen from bovine liver, ≥85%
Sigma-Aldrich
Glycogen from Mytilus edulis (Blue mussel), for DNA precipitations
Sigma-Aldrich
Glycogen from oyster, ≥75% dry basis
Sigma-Aldrich
N-(Phosphonomethyl)glycine, 96%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycogen from rabbit liver, ≥85% dry basis (enzymatic)
Sigma-Aldrich
Glycogen from Mytilus edulis (Blue mussel), ≥85%
Sigma-Aldrich
Isocitric Dehydrogenase (NADP) from porcine heart, Type IV, buffered aqueous glycerol solution, 3-20 units/mg protein
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, 99%, FCC