  • Search Results
  • Newly identified Chinese hamster ovary cell mutants defective in peroxisome biogenesis represent two novel complementation groups in mammals.
Newly identified Chinese hamster ovary cell mutants defective in peroxisome biogenesis represent two novel complementation groups in mammals.
European journal of cell biology (1997-08-01)
K Tateishi, K Okumoto, N Shimozawa, T Tsukamoto, T Osumi, Y Suzuki, N Kondo, I Okano, Y Fujiki
ABSTRACT
We isolated peroxisome biogenesis mutants from Chinese hamster ovary (CHO) cells, using the 9-(1'-pyrene)nonanol/ultraviolet (P9OH/ UV) method and wild-type CHO-K1 cells that had been stably transfected with cDNA encoding Pex2p (formerly peroxisome assembly factor-1, PAF-1). Three mutant cell clones, ZP110, ZP111, and ZP114, showed cytosolic localization of catalase, thereby indicating a defect in peroxisome biogenesis, whereas ZP112 and ZP113 contained fewer but larger catalase-positive particles. Mutant ZP115 displayed an aberrant, tubular structure immunoreactive to anti-catalase antibody. Mutants lacking morphologically recognizable peroxisomes also showed the typical peroxisome assembly-defective phenotype such as severe loss of catalase latency and resistance to 12-(1'-pyrene)dodecanoic acid (P12)/UV treatment. ZP110 and ZP111, and ZP114 were found to belong to two novel complementation groups, respectively, by complementation group analysis with cDNA transfection and cell fusion. Cell fusion with fibroblasts from patients with peroxisome biogenesis disorders such as Zellweger syndrome revealed that ZP110 and ZP114 could not be classified to any of human complementation groups. Thus, ZP110/ZP111 and ZP114 are the first, two peroxisome-deficient cell mutants of newly identified complementation groups distinct from the ten mammalian groups previously characterized.
Sigma-Aldrich
1-Pyrenedodecanoic acid, suitable for fluorescence, ≥98.0% (HPLC)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.