beta = 11.6 h. The bioavailability was about 52.8%. After ig [3H]ZA (4.44 MBq.kg-1), higher radioactivities were observed in kidney, liver, and gastrointestinal tract but lower in brain, fat, and femur. ZA given i.g. in rats was absorbed rapidly and distributed widely to most of the organs and tissues.