  • Search Results
  • Neurotoxicity of acetaldehyde-derived advanced glycation end products for cultured cortical neurons.
Neurotoxicity of acetaldehyde-derived advanced glycation end products for cultured cortical neurons.
Journal of neuropathology and experimental neurology (2003-05-29)
Masayoshi Takeuchi, Takayuki Watai, Nobuyuki Sasaki, Hiroshi Choei, Mina Iwaki, Takeshi Ashizawa, Yosuke Inagaki, Sho-Ichi Yamagishi, Seiji Kikuchi, Peter Riederer, Toshikazu Saito, Richard Bucala, Yukihiko Kameda
ABSTRACT
The Maillard reaction that leads to the formation of advanced glycation end products (AGEs) plays an important role in the pathogenesis of angiopathy in diabetic patients, in aging, and in neurodegenerative processes. We hypothesize that acetaldehyde (AA), one of the main metabolites of alcohol, may be involved in alcohol-induced neurotoxicity in vivo by formation of AA-derived AGEs (AA-AGEs) with brain proteins. Incubation of cortical neurons with AA-AGE produced a dose-dependent increase in neuronal cell-death, and the neurotoxicity of AA-AGE was neutralized by the addition of an anti-AA-AGE-specific antibody, but not by anti-N-ethyllysine (NEL) antibody. The AA-AGE epitope was detected in human brain of alcoholism. We propose that the structural epitope AA-AGE is an important toxic moiety for neuronal cells in alcoholism.
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Supelco
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Supelco
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, natural, FG
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde, ≥99%, meets FCC analytical specification
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol