  • Search Results
  • High-performance liquid chromatographic determination of nicotinamide and its metabolites in human and murine plasma and urine.
High-performance liquid chromatographic determination of nicotinamide and its metabolites in human and murine plasma and urine.
Journal of chromatography (1992-11-06)
M R Stratford, M F Dennis
ABSTRACT
A high-performance liquid chromatographic method is described which enables the determination of nicotinamide and eight of its possible metabolites in human and murine plasma and urine, using ion-pairing on a base-deactivated reversed-phase column. Calibration curves were linear up to 2 mumol/ml for nicotinamide and 200 nmol/ml for the metabolites; both the intra- and inter-assay relative standard deviations ranged between 1 and 8%. In murine plasma, the N-oxide was the major nicotinamide metabolite, but in man, formation of 1-methylnicotinamide and the 2- and 4-pyridones was also significant. In urine, nicotinuric acid was seen in the mouse, but no nicotinic acid metabolites were seen in man.
Product Description
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Nicotinamide (Niacinamide), analytical standard