  • Search Results
  • Human immunodeficiency virus type 1 Vpr induces the degradation of the UNG and SMUG uracil-DNA glycosylases.
Human immunodeficiency virus type 1 Vpr induces the degradation of the UNG and SMUG uracil-DNA glycosylases.
Journal of virology (2005-08-17)
Bärbel Schröfelbauer, Qin Yu, Samantha G Zeitlin, Nathaniel R Landau
ABSTRACT
The human immunodeficiency virus type 1 (HIV-1) accessory protein Vpr has previously been shown to bind to the cellular uracil DNA glycosylase UNG. We show here that the binding of Vpr to UNG and to the related enzyme SMUG induces their proteasomal degradation. UNG and SMUG were found to be encapsidated in Deltavpr HIV-1 virions but were significantly less abundant in vpr(+) virions. Deltavpr virions contained readily detectable uracil-DNA glycosylase enzymatic activity, while the activity was reduced to undetectable levels in vpr(+) virions. Consistent with proteasomal degradation, complexes that contained Vpr and the E3 ubiquitin ligase components Cul1 and Cul4 were detected in cell lysates. We hypothesized that the interaction of Vpr might be a means for the virus to reduce the frequency of abasic sites in viral reverse transcripts at uracil residues caused by APOBEC3-catalyzed deamination of cytosine residues. Although APOBEC3 is largely neutralized by the Vif accessory protein, residual enzyme could remain in virions that would generate uracils. In support of this, Deltavif vpr(+) HIV-1 produced in the presence of limited amounts of APOBEC3G was significantly more infectious than Deltavif Deltavpr virus. In Addition, vpr(+) HIV-1 replicated more efficiently than vpr(-) virus in cells that expressed limited amounts of APOBEC3G. The findings highlight the importance of cytidine deamination in the virus replication cycle and present a novel function for Vpr.
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Cytidine, 99%