  • Search Results
  • Simultaneous high performance liquid chromatographic separation of purines, pyrimidines, N-acetylated amino acids, and dicarboxylic acids for the chemical diagnosis of inborn errors of metabolism.
Simultaneous high performance liquid chromatographic separation of purines, pyrimidines, N-acetylated amino acids, and dicarboxylic acids for the chemical diagnosis of inborn errors of metabolism.
Clinical biochemistry (2005-09-06)
Barbara Tavazzi, Giuseppe Lazzarino, Paola Leone, Angela Maria Amorini, Francesco Bellia, Christopher G Janson, Valentina Di Pietro, Lia Ceccarelli, Sonia Donzelli, Jeremy S Francis, Bruno Giardina
ABSTRACT
To set up a novel simple, sensitive, and reliable ion-pairing HPLC method for the synchronous separation of several purines, pyrimidines, N-acetylated amino acids, and dicarboxylic acids for the chemical diagnosis and screening of inborn errors of metabolism (IEM). The separation was set up using a Hypersil C-18, 5-microm particle size, 250 x 4.6 mm column, and a step gradient using two buffers and tetrabutylammonium hydroxide as the pairing reagent. A highly sensitive diode array UV detector was set up at a wavelength between 200 and 300 nm that revealed purines and pyrimidines at 260 nm and other compounds at 206 nm. Compounds were determined in the plasma of 15 healthy adults, in the urine of 50 healthy subjects (1-3 years, 4-6 years, 8-10 years, 12-18 years, 25-35 years), and in 10 non-pathological amniotic fluid samples. To assess the validity of the chemical diagnosis of IEM, plasma and urine samples were analyzed in patients affected by Canavan disease (n = 10; mean age 4.6 +/- 2.3). Low plasma levels of N-acetylaspartate (16.96 +/- 19.57 micromol/L plasma; not detectable in healthy adults) and dramatically high urinary N-acetylaspartate concentrations (1872.03 +/- 631.86 micromol/mmol creatinine; 450 times higher than that which was observed in age-matched controls) were recorded. Neither N-acetylglutamate nor N-acetylaspartylglutamate could be detected in the plasma or urine of controls or patients with Canavan disease. The results demonstrate the suitability of the present ion-pairing HPLC separation with UV detection of cytosine, cytidine, creatinine, uracil, uridine, beta-pseudouridine, adenine, 3-methyladenine, hypoxanthine, xanthine, xanthosine, inosine, guanosine, ascorbic acid, thymine, thymidine, uric acid, 1-methyluric acid, orotic acid, N-acetylaspartate, N-acetylglutamate, N-acetylaspartylglutamate, malonic acid, methylmalonic acid, GSH, and GSSG as a reliable method for the prenatal and neonatal chemical diagnosis and screening of IEM using biological fluids.
Product Description
Supelco
Citric acid, certified reference material, TraceCERT®
Supelco
Succinic acid, certified reference material, TraceCERT®
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
L-Glutathione oxidized, ≥98% (HPLC)
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
L-Glutathione oxidized, ≥98%, lyophilized powder
Sigma-Aldrich
L-Glutathione oxidized, BioXtra, ≥98%
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Succinic acid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Methylmalonic acid, 99%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%
Supelco
Citric acid, analytical standard
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
N-Acetyl-Asp-Glu, ≥97% (TLC)
Sigma-Aldrich
Inosine, ≥99% (HPLC)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Supelco
Malonic acid, certified reference material, TraceCERT®
Sigma-Aldrich
N-Acetyl-L-glutamic acid, ReagentPlus®, 99%
Sigma-Aldrich
Malonic acid, ReagentPlus®, 99%
Sigma-Aldrich
N-Acetyl-L-aspartic acid, ≥99.0% (T)