  • Search Results
  • Effect of a rice bran fiber diet on serum glucose levels of diabetic patients in Brazil.
Effect of a rice bran fiber diet on serum glucose levels of diabetic patients in Brazil.
Archivos latinoamericanos de nutricion (2005-09-29)
Cecilia Rodrigues Silva, José Eduardo Dutra de Oliveira, Rui Augusto Hudari Gonçalves de Souza, Hugo Candido Silva
ABSTRACT
Eleven diabetic patients: 5, type 1 and 6, type 2 received a low-fiber diet (I) during 1 week and during the next 7 days the same diet, enriched with 40 g of fiber (30.6% insoluble and 11.7% soluble components) from rice bran (II) per day. Results showed that mean fasting and postprandial serum glucose levels were reduced, but values of high fiber diet were significantly lower (p < 0.001) than that of the lower fiber diet. For all patients, the high-fiber diet increased fecal weight. This increase was due to the fiber excreted, rather than water retained. There was no relationship between the increase in fiber intake and its fecal excretion. Sucrose and raffinose were found in the bran, but not in the feces. Lactose was present in the stools of the patients receiving enriched diet.
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20