  • Search Results
  • The B-vitamins in malnutrition with alcoholism. A model of intervitamin relationships.
The B-vitamins in malnutrition with alcoholism. A model of intervitamin relationships.
The British journal of nutrition (1976-09-01)
D K Dastur, N Santhadevi, E V Quadros, F C Avari, N H Wadia, M N Desai, E P Bharucha
PMID182198
ABSTRACT
1. The B-vitamin status of fifty-nine patients, mainly from the lower socio-economic classes in Bombay, with a history of chronic malnutrition, and of alcoholism of 1-5-20 years' duration, was studied before and during treatment, and in relation to their clinical, especially neurological, condition. These patients were divided into two neurological categories: (I) those with peripheral neuropathy (mainly sensory and distal) alone, (2) those with mental changes (mainly confusion and disorientation) also. Both categories frequently showed pellagrous pigmentation and mucocutaneous signs of B-vitamin deficiency. 2. Thiamin and erythrocyte transketolase (EC 2.2.1.1) activity, riboflavin, nicotinic acid, pantothenic acid, total and pyridoxal fraction of vitamin B6, folate and total vitamin B12 were estimated in the blood and the cerebrospinal fluid (CSF) of these patients, and also in the blood of sixty-nine control subjects and in the CSF of some of them. The concentrations of all the vitamins, except vitamin B12, were highly significantly lower in the blood of patients of category I compared to the controls, and erythrocyte transketolase activity and pyridoxal concentration in patients of category 2 were significantly lower than those of category I patients. Blood pantothenic acid and folate concentrations were reduced less consistently. 3. Serum vitamin B12 concentration was significantly increased (though within normal range) in the patients compared to the control group, probably because of the moderate hepatic insufficiency (as assessed by liver function tests) in the former. 4. The concentrations of thiamin, riboflavin, nicotinic acid and total vitamin B6 were also highly significantly lower in the CSF in patients of category I compared with controls. Furthermore, thiamin, nicotinic acid and total vitamin B6 concentrations were significantly lower in patients of category 2 than those of category I patients, indicating that CSF levels reflect better the neurological status of these patients. 5. There was a moderate increase in the blood concentration of all the vitamins tested, after a relatively poor hospital diet alone. There was a concurrent increase in the blood levels of thiamin, riboflavin, nicotinic acid and pantothenic acid after parenteral treatment with either thiamin or nicotinic acid. The administration of pyridoxine resulted in a significant increase in the blood levels of riboflavin and the pyridoxal fraction of vitamin B6.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Reagent Alcohol, reagent grade
Sigma-Aldrich
Ethyl alcohol, denatured, reagent grade
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG