  • Search Results
  • "Green" nanocomposites from cellulose acetate bioplastic and clay: effect of eco-friendly triethyl citrate plasticizer.
"Green" nanocomposites from cellulose acetate bioplastic and clay: effect of eco-friendly triethyl citrate plasticizer.
Biomacromolecules (2004-11-09)
Hwan-Man Park, Manjusri Misra, Lawrence T Drzal, Amar K Mohanty
ABSTRACT
"Green" nanocomposites have been successfully fabricated from cellulose acetate (CA) powder, eco-friendly triethyl citrate (TEC) plasticizer and organically modified clay. The effect of the amount of plasticizer varying from 15 to 40 wt % on the performance of the nanocomposites has been evaluated. The morphologies of these nanocomposites were evaluated through X-ray diffraction (XRD), atomic force microscopy (AFM), and transmission electron microscopy (TEM) studies. The mechanical properties of nanocomposites are correlated with the XRD and TEM observations. Cellulosic plastic-based nanocomposites with 20 wt % TEC plasticizer and 5 wt % organoclay showed better intercalation and an exfoliated structure than the counterpart having 30/40 wt % plasticizers. The tensile strength, modulus and thermal stability of cellulosic plastic reinforced with organoclay showed a decreasing trend with an increase of plasticizer content from 20 to 40 wt %. The nano-reinforcement at the lower volume fractions (phi < or = 0.02) reduced the water vapor permeability of cellulosic plastic by 2 times and the relative permeability better fits with larger platelet aspect ratios (alpha = 150).