  • Search Results
  • Two novel IL-1 family members, IL-1 delta and IL-1 epsilon, function as an antagonist and agonist of NF-kappa B activation through the orphan IL-1 receptor-related protein 2.
Two novel IL-1 family members, IL-1 delta and IL-1 epsilon, function as an antagonist and agonist of NF-kappa B activation through the orphan IL-1 receptor-related protein 2.
Journal of immunology (Baltimore, Md. : 1950) (2001-07-24)
R Debets, J C Timans, B Homey, S Zurawski, T R Sana, S Lo, J Wagner, G Edwards, T Clifford, S Menon, J F Bazan, R A Kastelein
ABSTRACT
IL-1 is of utmost importance in the host response to immunological challenges. We identified and functionally characterized two novel IL-1 ligands termed IL-1delta and IL-1epsilon. Northern blot analyses show that these IL-1s are highly abundant in embryonic tissue and tissues containing epithelial cells (i.e., skin, lung, and stomach). In extension, quantitative real-time PCR revealed that of human skin-derived cells, only keratinocytes but not fibroblasts, endothelial cells, or melanocytes express IL-1delta and epsilon. Levels of keratinocyte IL-1delta are approximately 10-fold higher than those of IL-1epsilon. In vitro stimulation of keratinocytes with IL-1beta/TNF-alpha significantly up-regulates the expression of IL-1epsilon mRNA, and to a lesser extent of IL-1delta mRNA. In NF-kappaB-luciferase reporter assays, we demonstrated that IL-1delta and epsilon proteins do not initiate a functional response via classical IL-1R pairs, which confer responsiveness to IL-1alpha and beta or IL-18. However, IL-1epsilon activates NF-kappaB through the orphan IL-1R-related protein 2 (IL-1Rrp2), whereas IL-1delta, which shows striking homology to IL-1 receptor antagonist, specifically and potently inhibits this IL-1epsilon response. In lesional psoriasis skin, characterized by chronic cutaneous inflammation, the mRNA expression of both IL-1 ligands as well as IL-1Rrp2 are increased relative to normal healthy skin. In total, IL-1delta and epsilon and IL-1Rrp2 may constitute an independent signaling system, analogous to IL-1alphabeta/receptor agonist and IL-1R1, that is present in epithelial barriers of our body and takes part in local inflammatory responses.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.