  • Search Results
  • Opposing activities of two novel members of the IL-1 ligand family regulate skin inflammation.
Opposing activities of two novel members of the IL-1 ligand family regulate skin inflammation.
The Journal of experimental medicine (2007-10-03)
Hal Blumberg, Huyen Dinh, Esther S Trueblood, James Pretorius, David Kugler, Ning Weng, Suzanne T Kanaly, Jennifer E Towne, Cynthia R Willis, Melanie K Kuechle, John E Sims, Jacques J Peschon
ABSTRACT
The interleukin (IL)-1 family members IL-1alpha, -1beta, and -18 are potent inflammatory cytokines whose activities are dependent on heterodimeric receptors of the IL-1R superfamily, and which are regulated by soluble antagonists. Recently, several new IL-1 family members have been identified. To determine the role of one of these family members in the skin, transgenic mice expressing IL1F6 in basal keratinocytes were generated. IL1F6 transgenic mice exhibit skin abnormalities that are dependent on IL-1Rrp2 and IL-1RAcP, which are two members of the IL-1R family. The skin phenotype is characterized by acanthosis, hyperkeratosis, the presence of a mixed inflammatory cell infiltrate, and increased cytokine and chemokine expression. Strikingly, the combination of the IL-1F6 transgene with an IL1F5 deficiency results in exacerbation of the skin phenotype, demonstrating that IL-1F5 has antagonistic activity in vivo. Skin from IL1F6 transgenic, IL1F5(-/-) pups contains intracorneal and intraepithelial pustules, nucleated corneocytes, and dilated superficial dermal blood vessels. Additionally, expression of IL1RL2, -1F5, and -1F6 is increased in human psoriatic skin. In summary, dysregulated expression of novel agonistic and antagonistic IL-1 family member ligands can promote cutaneous inflammation, revealing potential novel targets for the treatment of inflammatory skin disorders.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.