Enantioselective synthesis of dihydro-1H-benzindoles.
The Journal of organic chemistry (2013-03-14)
Gustavo P Silveira, Joseph P Marino
ABSTRACT
The first examples of dihydro-1H-benzindoles by enantioselective γ-lactamization reaction of naphthyl sulfilimines with trichloroacetyl chloride in the presence of ZnCu as catalyst (≥98:2 er and 65-80% yields) are described. Products are obtained by [3,3]-sigmatropic rearrangement of the azasulfonium enolate or followed by a second allylic rearrangement that transfers chirality. The absolute stereochemistry was confirmed by X-ray crystallography, which provides support for the mechanisms proposed.
Sigma-Aldrich
Trichloroacetyl chloride, 99%
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.