  • Search Results
  • Assessment of RNA integrity in the postmortem pig colonic tissue ex vivo.
Assessment of RNA integrity in the postmortem pig colonic tissue ex vivo.
Journal of animal science (2013-02-13)
B Bahar, J V O'Doherty, T Sweeney
ABSTRACT
Surgical removal of porcine intestinal tissue followed by an ex vivo challenge is an alternative technique of testing the anti-inflammatory effect of bioactive compounds in the intestine of live pigs. We investigated the effects of ex vivo incubation of porcine colonic tissue on the quantity and quality of total RNA over a 12-h time period. Colonic tissue of pig (n = 6) was surgically removed immediately postslaughter and incubated for 0, 3, 6, and 12 h in a humidified cell culture incubator with 5% CO(2) at 37°C. Tissue samples were processed for RNA extraction. The quantity and quality of total RNA were assessed on a NanoDrop Spectrophotometer and an Agilent 2100 Bioanalyzer, respectively. Ex vivo incubation had an effect on both the quantity (P < 0.001) and quality (P < 0.001) of total RNA. Relative to the RNA yield at 0 h (505.0 ± 48.64 μg/mg), the yield was significantly reduced after 6 h (227.6 ± 25.52 μg/mg; P < 0.001) and 12 h (159.3 ± 24.19 μg/mg; P < 0.001) of incubation. The 28S and 18S rRNA bands were visibly intact after 0, 3, and 6 h of incubation. However, after 12 h of incubation, a degraded RNA profile was evident. The RNA integrity number (RIN) values for the 0, 3, 6, and 12 h of incubation were 9.4 ± 0.10, 9.0 ± 0.10, 6.7 ± 0.17 (P < 0.001), and 3.3 ± 0.24 (P < 0.001), respectively. The transcript abundances of 4 constitutively expressed genes glyceraldehyde 3-phosphate dehydrogenase (GAPDH), beta actin (ACTB), beta 2-microglobulin (B2M), and peptidylprolyl isomerase A (PPIA) were reduced at both 6 and 12 h of incubation. It is concluded that ex vivo incubation of porcine colonic tissue up to 3 h postmortem generates good quality total RNA suitable for gene expression studies.
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 70 purifications
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 350 purifications
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 10 purifications
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.