  • Search Results
  • Characterization of a D-psicose-producing enzyme, D-psicose 3-epimerase, from Clostridium sp.
Characterization of a D-psicose-producing enzyme, D-psicose 3-epimerase, from Clostridium sp.
Biotechnology letters (2013-05-11)
Wanmeng Mu, Wenli Zhang, Dan Fang, Leon Zhou, Bo Jiang, Tao Zhang
ABSTRACT
The gene coding for D-psicose 3-epimerase (DPEase) from Clostridium sp. BNL1100 was cloned and expressed in Escherichia coli. The recombinant enzyme was purified by Ni-affinity chromatography. It was a metal-dependent enzyme and required Co(2+) as optimum cofactor. It displayed catalytic activity maximally at pH 8.0 and 65 °C (as measured over 5 min). The optimum substrate was D-psicose, and the K m, turnover number (k cat), and catalytic efficiency (k cat/K m) for D-psicose were 227 mM, 32,185 min(-1), and 141 min(-1 )mM(-1), respectively. At pH 8.0 and 55 °C, 120 g D-psicose l(-1) was produced from 500 g D-fructose l(-1) after 5 h.
Sigma-Aldrich
D-Psicose, ≥95%