  • Search Results
  • Visible-light-mediated Sr-Bi2O3 photocatalysis of tetracycline: kinetics, mechanisms and toxicity assessment.
Visible-light-mediated Sr-Bi2O3 photocatalysis of tetracycline: kinetics, mechanisms and toxicity assessment.
Chemosphere (2013-05-28)
Junfeng Niu, Shiyuan Ding, Liwen Zhang, Jinbo Zhao, Chenghong Feng
ABSTRACT
Photodegradation of tetracycline (TC) was investigated in aqueous solution by visible-light-driven photocatalyst Sr-doped β-Bi2O3 (Sr-Bi2O3) prepared via solvothermal synthesis. The decomposition of TC by Sr-Bi2O3 under visible light (λ>420nm) irradiation followed pseudo-first-order kinetics, and the removal ratio reached 91.2% after 120min of irradiation. Sr-Bi2O3 photocatalysis is able to break the naphthol ring of TC which decomposes to m-cresol via dislodging hydroxyl group step by step by photogenerated electron. This mechanism was verified by electron spin resonance measurement, the addition of radical scavengers and the intermediate product analysis, indicating that the photogenerated electron acts as a reductant and can be the key to the degradation process. In contrast, in TiO2 photocatalysis the naphthol ring is broken via oxidation by hydroxyl radical, while in direct photolysis the ring remains intact. In addition, the toxicity of photodegradation products was analyzed by bioluminescence inhibition. After 120min of irradiation by Sr-Bi2O3, the toxicity decreases by 90.6%, which is more substantial than direct photolysis (70%) and TiO2 photocatalysis (80%), indicating that the Sr-Bi2O3 photocatalysis is more eco-friendly than the other two methods.
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Bismuth(III) oxide, ReagentPlus®, powder, 10 μm, 99.9% trace metals basis
Sigma-Aldrich
Tetracycline hydrochloride, powder
Sigma-Aldrich
Bismuth(III) oxide, powder, 99.999% trace metals basis
Supelco
Tetracycline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetracycline hydrochloride, VETRANAL®, analytical standard
Sigma-Aldrich
Bismuth, powder, −100 mesh, 99% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Bismuth, granular, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
Sigma-Aldrich
Bismuth(III) oxide, nanopowder, 90-210 nm particle size, 99.8% trace metals basis
Sigma-Aldrich
Strontium, granular, 99% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Bismuth(III) oxide, purum, ≥98.0% (KT)
Sigma-Aldrich
Bismuth, shot, 4-30 mesh, 99.9% trace metals basis
Sigma-Aldrich
Strontium, random pieces, 99%
Sigma-Aldrich
Bismuth, beads, 1-5 mm, 99.999% trace metals basis