  • Search Results
  • Effects of cooking methods and chemical tenderizers on survival of Escherichia coli O157:H7 in ground beef patties.
Effects of cooking methods and chemical tenderizers on survival of Escherichia coli O157:H7 in ground beef patties.
Meat science (2013-06-12)
Yohan Yoon, Ifigenia Geornaras, Avik Mukherjee, Keith E Belk, John A Scanga, Gary C Smith, John N Sofos
ABSTRACT
This study evaluated chemical tenderizers and cooking methods to inactivate Escherichia coli O157:H7 in ground beef patties (model system for non-intact beef). Ground beef was inoculated with E. coli O157:H7 and mixed with (i) nothing (control), (ii) calcium chloride (CC) and flavoring agents (FA), (iii) CC, FA, and acetic acid (AA), (iv) sodium chloride (SC), sodium tripolyphosphate (ST), and potassium lactate (PL), and (v) the combination of SC, ST, PL, and AA. Patties were stored in aerobic or vacuum bags at -20, 4, and 12°C. Samples were grilled, broiled, or pan-fried to 60 or 65°C. Total bacterial and E. coli O157:H7 populations remained unchanged during storage. Broiling was more effective in reducing E. coli O157:H7 than grilling and pan-frying, and acidified tenderizers reduced E. coli O157:H7 more than non-acidified tenderizers in broiling. Higher reductions were observed at 65°C than 60°C in broiled and grilled samples. These results indicate that acidified tenderizers and broiling may be useful in non-intact beef safety.
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Sodium tripolyphosphate, technical grade, 85%
Sigma-Aldrich
Sodium triphosphate pentabasic, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Calcium chloride dihydrate, puriss., meets analytical specification of Ph. Eur., USP, FCC, E509, 99-103%, ≤0.0001% Al
Sigma-Aldrich
Calcium chloride dihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, for molecular biology, ≥99.0%
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)