  • Search Results
  • Comparison of the effect of ranibizumab and verteporfin for polypoidal choroidal vasculopathy: 12-month LAPTOP study results.
Comparison of the effect of ranibizumab and verteporfin for polypoidal choroidal vasculopathy: 12-month LAPTOP study results.
American journal of ophthalmology (2013-07-24)
Akio Oishi, Hiroshi Kojima, Michiko Mandai, Shigeru Honda, Toshiyuki Matsuoka, Hideyasu Oh, Mihori Kita, Tomoko Nagai, Masashi Fujihara, Nobuhiro Bessho, Mamoru Uenishi, Yasuo Kurimoto, Akira Negi
ABSTRACT
To compare the effect of photodynamic therapy (PDT) and intravitreal ranibizumab in patients with polypoidal choroidal vasculopathy (PCV). Randomized clinical trial. Multicenter. Total of 93 patients with treatment-naïve PCV. Patients were randomized to 2 arms. Patients in the PDT arm underwent a single session of PDT with verteporfin, and patients in the ranibizumab arm received 3 monthly ranibizumab injections at baseline. Additional treatment was performed as needed in each arm. Primary outcome measurement was the proportion of patients gaining or losing more than 0.2 logarithm of minimal angle of resolution (logMAR) units from baseline. Mean change of logMAR and central retinal thickness (CRT) were also evaluated. In the PDT arm (n = 47), 17.0% achieved visual acuity gain, 55.3% had no change, and 27.7% experienced visual acuity loss. The results were 30.4%, 60.9%, and 8.7%, respectively, in the ranibizumab arm (n = 46), significantly better than the PDT arm (P = .039). In the PDT arm, mean CRT improved (366.8 ± 113.6 μm to 289.1 ± 202.3 μm, P < .001), but logMAR was unchanged (0.57 ± 0.31 to 0.62 ± 0.40). The ranibizumab arm demonstrated improvement in both CRT (418.9 ± 168.6 μm to 311.2 ± 146.9 μm, P < .001) and logMAR (0.48 ± 0.27 to 0.39 ± 0.26, P = .003). Mean change of logMAR was also greater in the ranibizumab arm (P = .011). Intravitreal injection of ranibizumab is more effective than PDT for treatment-naïve PCV.
Sigma-Aldrich
Verteporfin, ≥94% (HPLC)