  • Search Results
  • Ectopic calcification in β-thalassemia patients is associated with increased oxidative stress and lower MGP carboxylation.
Ectopic calcification in β-thalassemia patients is associated with increased oxidative stress and lower MGP carboxylation.
Biochimica et biophysica acta (2013-08-01)
Federica Boraldi, Maria Garcia-Fernandez, Chiara Paolinelli-Devincenzi, Giulia Annovi, Leon Schurgers, Cees Vermeer, Paolo Cianciulli, Ivonne Ronchetti, Daniela Quaglino
ABSTRACT
A number of beta-thalassemia (β-thal) patients in the course of the disease exhibit ectopic calcification affecting skin, eyes and the cardiovascular system. Clinical and histopathological features have been described similar to those in pseudoxanthoma elasticum (PXE), although different genes are affected in the two diseases. Cultured dermal fibroblasts from β-thal patients with and without PXE-like clinical manifestations have been compared for parameters of redox balance and for the expression of proteins, which have been already associated with the pathologic mineralisation of soft connective tissues. Even though oxidative stress is a well-known condition of β-thal patients, our results indicate that the occurrence of mineralized elastin is associated with a more pronounced redox disequilibrium, as demonstrated by the intracellular increase of anion superoxide and of oxidized proteins and lipids. Moreover, fibroblasts from β-thal PXE-like patients are characterized by decreased availability of carboxylated matrix Gla protein (MGP), as well as by altered expression of proteins involved in the vitamin K-dependent carboxylation process. Results demonstrate that elastic fibre calcification is promoted when redox balance threshold levels are exceeded and the vitamin K-dependent carboxylation process is affected decreasing the activity of MGP, a well-known inhibitor of ectopic calcification. Furthermore, independently from the primary gene defect, these pathways are similarly involved in fibroblasts from PXE and from β-thal PXE-like patients as well as in other diseases leading to ectopic calcification, thus suggesting that can be used as markers of pathologic mineralisation.
Sigma-Aldrich
Elastin from bovine neck ligament, powder
Sigma-Aldrich
Elastin, soluble from bovine neck ligament, salt-free, lyophilized powder
Sigma-Aldrich
Elastin from human skin, insoluble powder
Sigma-Aldrich
Elastin, soluble from human lung, lyophilized powder
Sigma-Aldrich
Elastin, soluble from human aorta, lyophilized powder
Sigma-Aldrich
Elastin from mouse lung, powder