  • Search Results
  • Micromonospora kangleipakensis sp. nov., isolated from a sample of limestone quarry.
Micromonospora kangleipakensis sp. nov., isolated from a sample of limestone quarry.
International journal of systematic and evolutionary microbiology (2013-08-03)
Salam Nimaichand, Yong-Guang Zhang, Juan Cheng, Li Li, Dao-Feng Zhang, En-Min Zhou, Lei Dong, Debananda S Ningthoujam, Wen-Jun Li
ABSTRACT
Strain MBRL 34(T), isolated from a sample of limestone quarry located at Hundung, Manipur, India, was characterized by polyphasic taxonomy. The strain showed the highest 16S rRNA gene sequence similarity with Micromonospora echinaurantica DSM 43904(T) (98.4 %), but formed a monophyletic clade with Micromonospora coerulea DSM 43143(T) (98.3 %) in the neighbour-joining tree. DNA-DNA hybridization experiments gave a DNA-DNA relatedness value of 53.1 % between MBRL 34(T) and M. coerulea DSM 43143(T). Strain MBRL 34(T) contained meso-diaminopimelic acid, galactose and glucose in the whole-cell hydrolysates along with small amounts of mannose, xylose, rhamnose and ribose. The major polar lipids detected were diphosphatidylglycerol, phosphatidylethanolamine, phosphatidylglycerol, phosphatidylinositol and phosphatidylinositolmannoside, along with an unknown lipid. MK-10(H6), MK-10(H2) MK-11(H4) and MK-10(H4) were the predominant menaquinones detected. The major fatty acids were iso-C16 : 0 and iso-C15 : 0. The G+C content of the genomic DNA was 73.5 %. Based on the taxonomic characteristics from a polyphasic study, strain MBRL 34(T) merits recognition as a representative of a novel species of the genus Micromonospora for which the name Micromonospora kangleipakensis sp. nov. is proposed; the type strain is MBRL 34(T) ( = DSM 45612(T) = JCM 17696(T)).
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
2,6-Diaminopimelic acid, ≥98% (TLC)
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Supelco
Calcium carbonate, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
2,6-Diaminopimelic acid, ≥97.0% (NT)
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%