  • Search Results
  • Application of surfactant assisted dispersive liquid-liquid microextraction as an efficient sample treatment technique for preconcentration and trace detection of zonisamide and carbamazepine in urine and plasma samples.
Application of surfactant assisted dispersive liquid-liquid microextraction as an efficient sample treatment technique for preconcentration and trace detection of zonisamide and carbamazepine in urine and plasma samples.
Journal of chromatography. A (2013-08-21)
Mohammad Behbahani, Fatemeh Najafi, Saman Bagheri, Majid Kalate Bojdi, Mani Salarian, Akbar Bagheri
ABSTRACT
A simple, rapid, and efficient method, based on surfactant assisted dispersive liquid-liquid microextraction (SA-DLLME), followed by high performance liquid chromatography (HPLC) has been developed for simultaneous preconcentration and trace detection of zonisamide and carbamazepine in biological samples. A conventional cationic surfactant called cethyltrimethyl ammonium bromide (CTAB) was used as a disperser agent in the proposed approach. 1.5 mL of CTAB (0.45 mmol L(-1)) (disperser solvent) containing 50.0 μL of 1-octanol (extraction solvent) was injected rapidly into the 7.0 mL of water or diluted plasma or urine. A cloudy solution (water, 1-octanol, and CTAB) was formed in the test tube. After formation of cloudy solution, the mixture was centrifuged and 20 μL of collected phase was injected into HPLC for subsequent analysis. Some parameters such as the type and volume of the extraction solvent, the type and concentration of surfactant, pH, ionic strength and centrifugation time were evaluated and optimized. Under optimum extraction conditions, the limits of detections (LODs) were 2.1 and 1.5 μg L(-1) (based on 3Sb/m) for urine samples, and 2.3 and 1.6 μg L(-1) for plasma samples. Linear dynamic range of 5-300 and 5-200 μg L(-1) were obtained for zonisamide and carbamazepine in all samples. Finally, the applicability of the proposed method was evaluated by extraction and determination of the drugs in urine and plasma samples.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Supelco
1-Octanol, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Sigma-Aldrich
Cetyltrimethylammonium hydrogensulfate, 99%
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
1-Octanol, ACS reagent, ≥99%
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
1-Octanol, suitable for HPLC, ≥99%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%