  • Search Results
  • A new method for pH triggered curcumin release by applying poly(L-lysine) mediated nanoparticle-congregation.
A new method for pH triggered curcumin release by applying poly(L-lysine) mediated nanoparticle-congregation.
Analytica chimica acta (2013-09-04)
Digambara Patra, Fatima Sleem
ABSTRACT
We introduce a novel method for encapsulation of curcumin by synthesizing microcapsule containing self-assembled nanoparticles using poly (L-lysine), trisodium citrate and silica sol. Such microcapsules can only be prepared in neutral and alkaline environment and unencapsulated curcumin can be effectively removed by simple centrifugation with encapsulation efficiency 57.34%. The particle sizes are in the range 0.7-3 μm with an effective diameter 1.05 μm. The structure of the microcapsules is dependent upon the solubility of curcumin in the solvent environment, the microcapsule are spherical when prepared in 10% acetone and bowl-shaped/conical when prepared in water suspension, however, the size of these curcumin encapsulated microcapsules remain similar. Fluorescence microscope images confirm that curcumin is predominantly concentrated within the shell wall of the capsules. Photophysical behavior of Micro-curcumin with respect to curcumin alone is evaluated. Release of curcumin is found to be triggered by pH where acidic environment trigger maximum release compared to basic and neutral conditions. Micro-curcumin is as stable as curcumin. Drug release efficiency is found to be 61.44% and the drug release profile of Micro-curcumin follow Higuchi model. It is also revealed that β-diketone group of curcumin responsible for scavenging activity is retained in the Micro-curcumin, thus suggesting applicability of such system as a poorly water soluble drug delivery vehicle. In contrast to other curcumin delivery systems, the presented method is easy, fast and does not need flow rate monitoring device. In addition poly (L-lysine) as a non-toxic and highly stable material that prevents metabolic degradation is used in the present preparation method.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Supelco
Acetone, analytical standard
Supelco
Curcumin, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Curcumin, primary reference standard
Supelco
Acetone solution, contains 20.0 % (v/v) acetonitrile, suitable for HPLC