  • Search Results
  • Iron-catalyzed homocoupling of aryl halides and derivatives in the presence of alkyllithiums.
Iron-catalyzed homocoupling of aryl halides and derivatives in the presence of alkyllithiums.
Organic letters (2013-09-06)
Dounia Toummini, Fouad Ouazzani, Marc Taillefer
ABSTRACT
Direct synthesis of biaryl derivatives from aryl halides takes place under very mild temperature conditions by using a ligand-free iron catalytic system. The procedure, which proceeds via an in situ quantitative aryl halide exchange with alkyllithiums, allows for excellent control of the reactivity and is in line with the sustainable development. The method is also applicable to styryl and benzyl halides and to phenylacetylene.
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Lithium, granular, 99% trace metals basis