  • Search Results
  • The neutrophil elastase inhibitor sivelestat suppresses accelerated gastrointestinal tumor growth via peritonitis after cecal ligation and puncture.
The neutrophil elastase inhibitor sivelestat suppresses accelerated gastrointestinal tumor growth via peritonitis after cecal ligation and puncture.
Anticancer research (2013-09-12)
Koshi Kumagai, Yoshiro Saikawa, Hiroya Takeuchi, Koichi Suda, Kazumasa Fukuda, Rieko Nakamura, Tsunehiro Takahashi, Hirofumi Kawakubo, Norihito Wada, Taku Miyasho, Yuko Kitagawa
ABSTRACT
We examined whether tumor growth is enhanced by cecal ligation and puncture (CLP) and suppressed by a neutrophil elastase inhibitor, sivelestat. C57BL/6 mice were divided in CLP/sivelestat, CLP alone, and control (simple laparotomy) groups. Murine CT26 colon carcinoma cells were injected subcutaneously into the back of each mouse and tumor growth and serum cytokine levels were assessed. Mice subjected to CLP alone exhibited enhanced tumor growth compared to controls with subcutaneously injected CT26 cells (0.64±0.24 g vs. 0.021±0.027 g, p<0.001), while treatment with CLP/sivelestat produced smaller tumors than CLP alone (0.28±0.23 g vs. 0.64±0.24 g, p=0.006). Cytokine assays showed suppressed production of interleukin (IL)-6 and IL-10 in the CLP/sivelestat group, and increased IL-6 and IL-10 in the CLP-alone group. Intra-abdominal inflammation induced by CLP enhances the growth of subcutaneously implanted tumors, while perioperative administration of sivelestat suppresses tumor growth by affecting systemic inflammation.
Sigma-Aldrich
Elastase from human leukocytes, lyophilized powder, ≥50 units/mg protein (Bradford)
Sigma-Aldrich
Sivelestat sodium salt hydrate, ≥98% (HPLC), solid