Elastin staining patterns in primary cicatricial alopecia.
Journal of the American Academy of Dermatology (2013-09-17)
Maxwell A Fung, Victoria R Sharon, Mondhipa Ratnarathorn, Thomas H Konia, Keira L Barr, Paradi Mirmirani
ABSTRACT
Most biopsy specimens of cicatricial (scarring) alopecia can be readily subclassified as lymphocytic versus neutrophilic, but specific diagnosis remains difficult, particularly when a late stage of the disease is sampled. We sought to document patterns of scarring highlighted by elastic tissue staining in primary cicatricial alopecia. We documented Verhoeff elastic van Gieson staining patterns in 58 routinely embedded (vertical) biopsy specimens of cicatricial alopecia. Patterns of fibrosis included perifollicular (wedge-shaped vs broad tree trunk-shaped) and diffuse. The patterns were compared against the diagnosis obtained by independent expert clinical review, including central centrifugal cicatricial alopecia (CCCA), lichen planopilaris, traction alopecia, frontal fibrosing alopecia, discoid lupus erythematosus, and tufted folliculitis. Wedge-shaped perifollicular fibrosis was seen in lichen planopilaris but also in CCCA. Broad tree trunk-shaped perifollicular fibrosis was most commonly encountered in CCCA. The retrospective nature of the study precluded temporal staging of the disease process. Patterns of fibrosis highlighted by elastin staining in primary cicatricial alopecia appear to be disease specific. Superficial wedge-shaped perifollicular fibrosis is associated with but may not be specific for lichen planopilaris. Broad tree trunk-like perifollicular fibrosis is specific for CCCA but not present in many cases. Elastin staining represents a useful ancillary study for the evaluation of late-stage scarring alopecia in routinely oriented punch biopsy specimens.
Sigma-Aldrich
Elastin from bovine neck ligament, powder
Sigma-Aldrich
Elastin, soluble from bovine neck ligament, salt-free, lyophilized powder
Sigma-Aldrich
Elastin from human skin, insoluble powder
Sigma-Aldrich
Elastin, soluble from human lung, lyophilized powder
Sigma-Aldrich
Elastin, soluble from human aorta, lyophilized powder
Sigma-Aldrich
Elastin from mouse lung, powder