  • Search Results
  • Synergistic role of different soil components in slow sorption kinetics of polar organic contaminants.
Synergistic role of different soil components in slow sorption kinetics of polar organic contaminants.
Environmental pollution (Barking, Essex : 1987) (2013-09-21)
Dongmei Zhang, Lei Hou, Dongqiang Zhu, Wei Chen
ABSTRACT
We observed that the sorption kinetics of nitrobenzene and 2,4-dinitrotoluene (two model polar compounds) was significantly slower than that of 1,4-dichlorobenzene and phenanthrene (two model apolar compounds). The difference was attributable to the strong non-hydrophobic interactions between the polar molecules and soil. Interestingly, sorption kinetics of the polar sorbates to the soil organic matter-free soil, humic/fulvic acid-free soil, and extracted humic acids was very fast, indicating that different soil components played a synergetic role in the observed slow kinetics. We propose that slow sorption kinetics of highly polar sorbates stems mainly from the strong specific interactions (H-bonding, electron donor-acceptor interactions, etc.) with humic/fulvic acids; such specific interactions occur when sorbate molecules diffuse through humic/fulvic acids coiled, in relatively compressed confirmations, within the complex, tortuous, and porous soil matrices formed by mineral grains/particles and soil organic matter.
Product Description
Supelco
Phenanthrene solution, certified reference material, 5000 μg/mL in methanol
Supelco
2,4-Dinitrotoluene, analytical standard
Sigma-Aldrich
2,4-Dinitrotoluene, 97%
Supelco
1,4-Dichlorobenzene solution, certified reference material, 5000 μg/mL in methanol
Supelco
2,4-Dinitrotoluene solution, 10 mg/mL in acetonitrile, ampule of 5 mL, certified reference material, Cerilliant®
Sigma-Aldrich
1,4-Dichlorobenzene, ≥99%
Sigma-Aldrich
Phenanthrene, 98%
Supelco
1,4-Dichlorobenzene, PESTANAL®, analytical standard
Supelco
Nitrobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
Nitrobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Nitrobenzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenanthrene, sublimed grade, ≥99.5%
Supelco
Phenanthrene, analytical standard, for environmental analysis
Supelco
Nitrobenzene solution, certified reference material, 5000 μg/mL in methanol
Supelco
Phenanthrene, certified reference material, TraceCERT®