  • Search Results
  • Persistent sodium current drives conditional pacemaking in CA1 pyramidal neurons under muscarinic stimulation.
Persistent sodium current drives conditional pacemaking in CA1 pyramidal neurons under muscarinic stimulation.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-09-21)
Jason Yamada-Hanff, Bruce P Bean
ABSTRACT
Hippocampal CA1 pyramidal neurons are normally quiescent but can fire spontaneously when stimulated by muscarinic agonists. In brain slice recordings from mouse CA1 pyramidal neurons, we examined the ionic basis of this activity using interleaved current-clamp and voltage-clamp experiments. Both in control and after muscarinic stimulation, the steady-state current-voltage curve was dominated by inward TTX-sensitive persistent sodium current (I(NaP)) that activated near -75 mV and increased steeply with depolarization. In control, total membrane current was net outward (hyperpolarizing) near -70 mV so that cells had a stable resting potential. Muscarinic stimulation activated a small nonselective cation current so that total membrane current near -70 mV shifted to become barely net inward (depolarizing). The small depolarization triggers regenerative activation of I(NaP), which then depolarizes the cell from -70 mV to spike threshold. We quantified the relative contributions of I(NaP), hyperpolarization-activated cation current (I(h)), and calcium current to pacemaking by using the cell's own firing as a voltage command along with specific blockers. TTX-sensitive sodium current was substantial throughout the entire interspike interval, increasing as the membrane potential approached threshold, while both Ih and calcium current were minimal. Thus, spontaneous activity is driven primarily by activation of I(NaP) in a positive feedback loop starting near -70 mV and providing increasing inward current to threshold. These results show that the pacemaking "engine" from I(NaP) is an inherent property of CA1 pyramidal neurons that can be engaged or disengaged by small shifts in net membrane current near -70 mV, as by muscarinic stimulation.
Sigma-Aldrich
Nickel(II) chloride, 98%
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC)
Sigma-Aldrich
Nickel(II) chloride hexahydrate, 99.9% trace metals basis
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel(II) chloride hexahydrate, ReagentPlus®
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
Nickel(II) chloride hexahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Nickel(II) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
DL-2-Amino-5-phosphonopentanoic acid, solid
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Acetylcholine chloride, suitable for cell culture
Sigma-Aldrich
Nickel(II) chloride hexahydrate, puriss. p.a., ≥98%
SAFC
L-Valine
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Acetylcholine iodide, ≥97%
Sigma-Aldrich
Nickel(II) chloride hexahydrate, 99.999% trace metals basis
Sigma-Aldrich
NBQX disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 4200, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
CGP 55845 hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis