  • Search Results
  • Optical, bactericidal and water repellent properties of electrospun nano-composite membranes of cellulose acetate and ZnO.
Optical, bactericidal and water repellent properties of electrospun nano-composite membranes of cellulose acetate and ZnO.
Carbohydrate polymers (2013-09-26)
S Anitha, B Brabu, D John Thiruvadigal, C Gopalakrishnan, T S Natarajan
ABSTRACT
In this report, ZnO nanoparticles embedded cellulose acetate (CA) fibrous membrane with multifunctional properties have been prepared through electrospinning method. The morphology of the electrospun composite membrane was analyzed by scanning electron microscope (SEM). It was found that the polymer concentration in the solution has a significant effect on the morphology of the fibers. The optical property of the sample was tested using photo luminescence (PL) spectra. There is no significant change in the emission features of cellulose acetate with the addition of ZnO. The anti-bacterial property of the sample was studied using disk diffusion method. The wettability of the pure and composite fibrous membrane was also studied by measuring the contact angle of water on the membrane. It was observed that the embedded ZnO in the CA was responsible for the hydrophobic nature of the surface.
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Cellulose acetate, average Mn ~50,000 by GPC
Sigma-Aldrich
Cellulose acetate, average Mn ~30,000 by GPC
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Zinc oxide, nanopowder, <100 nm particle size
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Supelco
Cellulose, acid washed, from spruce, for column chromatography
Supelco
Cellulose, powder, for column chromatography
Supelco
Cellulose, acid washed, powder, for column chromatography
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, fibers, (medium)
Sigma-Aldrich
Zinc oxide, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, 40 wt. % in ethanol, <130 nm particle size
Supelco
Zinc oxide, analytical standard