  • Search Results
  • A comparative study of antitumor activities and DNA cleavage on a class of dehydroabietylamine derivatives.
A comparative study of antitumor activities and DNA cleavage on a class of dehydroabietylamine derivatives.
Die Pharmazie (2014-01-02)
Chao-Xiang Liu, Zhong-Xiang Lin, Xing Yu, Zhou Lu, Ai-Min Zhou, Yong-Li Bao
ABSTRACT
A series of novel dehydroabietylamine derivatives containing tricyclic diterpene structures were synthesized. The antitumor activities of these compounds against L02, Hey-1B and HepG2 cells were investigated. Significant activity was discovered forfourteen analogs. Meanwhile these compounds exhibit DNA cleavage activities on plasmid DNA (Escherichia coli), which depend on the Schiff base structure and the substituent of the aromatic moiety. Our findings present further information on the relationship between the chemical structure, biological function and DNA cleavage characteristics.
Sigma-Aldrich
Iron, ≥99%, reduced, powder (fine)
Sigma-Aldrich
Carbonyl iron, ≥97% Fe basis
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Iron, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, granular, 10-40 mesh, >99.99% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
Iron, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Iron, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Iron, rod, diam. 6.3 mm, 99.98% trace metals basis
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, wire, diam. 0.5 mm, ≥99.9% trace metals basis