MilliporeSigma
[Endoscopic resection of intestinal mucosa for colorectal neoplasia].
Klinichna khirurhiia (2014-02-08)
V O Iakovenko, O H Kuryk
ABSTRACT
The results of endoscopic resection of intestinal mucosa for colorectal neoplasia were analyzed. There were excised 148 colorectal lesions in 116 patients.The operation was performed, using submucosal injection of adrenaline solution with indigocarmine and the loop resection technique. Endoscopic resection of intestinal mucosa together with pathologic lesion constitute a safe and effective method of treatment of colorectal neoplasia.