  • Search Results
  • Standardization of DNA extraction from methanol acetic acid fixed cytogenetic cells of cattle and buffalo.
Standardization of DNA extraction from methanol acetic acid fixed cytogenetic cells of cattle and buffalo.
Pakistan journal of biological sciences : PJBS (2014-02-11)
Rosaiah Kotikalapudi, Rajesh K Patel, Sanghamitra Katragadda
ABSTRACT
The aim of the study is to standardize the simple method for extracting DNA from cells fixed in fixative (3:1 ratio of methanol and acetic acid glacial) mostly used for chromosomal studies in cattle and buffaloes. These fixed cells were stored for more than 6 months at refrigerated temperature. The fixed cells were washed 2-3 times by the ice cold 1x Phosphate Buffer Saline (PBS) with pH 7.4, so that effect of fixative may be eliminated. The genomic DNA was extracted by adding cell lysis and nucleus lysis buffers. The quality and quantity of DNA were estimated. The readings of nano drop and agarose gel electrophoresis indicate good quality DNA isolated with a rapid and simple protocol routinely using in our laboratory. The method enables us to study the DNA of a cattle and buffaloes after completing cytogenetic investigation or in cases where DNA samples are otherwise not available. This protocol may be useful for molecular analysis of DNA from fixed cells palettes.
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Methanol solution, contains 0.10 % (v/v) trifluoroacetic acid, 10 % (v/v) water
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, NMR reference standard