  • Search Results
  • Removal of Cd2+ from water by Friedel's salt (FS: 3CaO x A12O3 x CaCI2 x 10H2O): sorption characteristics and mechanisms.
Removal of Cd2+ from water by Friedel's salt (FS: 3CaO x A12O3 x CaCI2 x 10H2O): sorption characteristics and mechanisms.
Journal of environmental sciences (China) (2014-02-14)
Juanjuan Zhang, He Zhao, Hongbin Cao, Heping Li, Zhibao Li
ABSTRACT
The development of low-cost and efficient new mineral adsorbents has been a hot topic in recent years. In this study, Friedel's salt (FS: 3CaO x A12O3 x CaCl2 x10H2O), a hexagonal layered inorganic absorbent, was synthesized to remove Cd2+ from water. The adsorption process was simulated by Langmuir and Freundlich models. The adsorption mechanism was further analyzed with TEM, XRD, FT-IR analysis and monitoring of metal cations released and solution pH variation. The results indicated the adsorbent FS had an outstanding ability for Cd(II) adsorption. The maximum adsorption capacity of the FS for Cd(II) removal can reach up to 671.14 mg/g. The nearly equal numbers of Cd2+ adsorbed and Ca2+ released demonstrated that ion-exchange (both surface and inner) of the FS for Cd(II) played an important role during the adsorption process. Furthermore, the surface of the FS after adsorption was microscopically disintegrated while the inner lamellar structure was almost unchanged. The behavior of Cd(II) adsorption by FS was significantly affected by surface reactions. The mechanisms of Cd2+ adsorption by the FS mainly included surface complexation and surface precipitation. In the present study, the adsorption process was fitted better by the Langmuir isotherm model (R2 = 0.9999) than the Freundlich isotherm model (R2 = 0.8122). Finally, due to the high capacity for ion-exchange on the FS surface, FS is a promising layered inorganic adsorbent for the removal of Cd(II) from water.
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Aluminum oxide, for the determination of hydrocarbons
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Cadmium, rod, diam. 4.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh