  • Search Results
  • Screening of rare earth metal grafted MCM-41 silica for asymmetric catalysis.
Screening of rare earth metal grafted MCM-41 silica for asymmetric catalysis.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association (2001)
Gerstberger G and Anwander R.