  • Search Results
  • Development of a simple and rapid solid phase microextraction-gas chromatography-triple quadrupole mass spectrometry method for the analysis of dopamine, serotonin and norepinephrine in human urine.
Development of a simple and rapid solid phase microextraction-gas chromatography-triple quadrupole mass spectrometry method for the analysis of dopamine, serotonin and norepinephrine in human urine.
Analytica chimica acta (2014-01-21)
Attilio Naccarato, Emanuela Gionfriddo, Giovanni Sindona, Antonio Tagarelli
ABSTRACT
The work aims at developing a simple and rapid method for the quantification of dopamine (DA), serotonin (5-HT) and norepinephrine (NE) in human urine. The urinary levels of these biogenic amines can be correlated with several pathological conditions concerning heart disease, stress, neurological disorders and cancerous tumors. The proposed analytical approach is based on the use of solid phase microextraction (SPME) combined with gas chromatography-triple quadrupole mass spectrometry (GC-QqQ-MS) after a fast derivatization of both aliphatic amino and phenolic moieties by propyl chloroformate. The variables influencing the derivatization reaction were reliably optimized by the multivariate approach of "Experimental design". The optimal conditions were obtained by performing derivatization with 100μL of propyl chloroformate and 100μL of pyridine. The extraction ability of five commercially available SPME fibers was evaluated in univariate mode and the best results were obtained using the polyacrylate fiber. The variables affecting the efficiency of SPME analysis were again optimized by the multivariate approach of "Experimental design" and, in particular, a central composite design (CCD) was applied. The optimal values were extraction in 45min at room temperature, desorption temperature at 300°C, no addition of NaCl. Assay of derivatized analytes was performed by using a gas chromatography-triple quadrupole mass spectrometry (GC-QqQ-MS) system in selected reaction monitoring (SRM) acquisition. An evaluation of all analytical parameters demonstrates that the developed method provides satisfactory results. Indeed, very good linearities were achieved in the tested calibration range with correlation coefficient values of 0.9995, 0.9999 and 0.9997 for DA, 5-HT and NE, respectively. Accuracies and RSDs calculated for between-run and tested at concentrations of 30, 200, and 800μg L(-1) were in the range from 92.8% to 103.0%, and from 0.67 to 4.5%, respectively. Finally, the LOD values obtained can be considered very good (0.587, 0.381 and 1.23μg L(-1) for DA, 5-HT and NE, respectively).
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Ethyl chloroformate, 97%
Supelco
Serotonin, analytical standard
Sigma-Aldrich
Methyl chloroformate, 99%
Sigma-Aldrich
DL-Norepinephrine hydrochloride, crystalline, ≥97% (TLC)
Sigma-Aldrich
Propyl chloroformate, 98%