  • Search Results
  • Equivocal colonic carcinogenicity of Aloe arborescens Miller var. natalensis berger at high-dose level in a Wistar Hannover rat 2-y study.
Equivocal colonic carcinogenicity of Aloe arborescens Miller var. natalensis berger at high-dose level in a Wistar Hannover rat 2-y study.
Journal of food science (2009-03-28)
M Yokohira, Y Matsuda, S Suzuki, K Hosokawa, K Yamakawa, N Hashimoto, K Saoo, K Nabae, Y Doi, T Kuno, K Imaida
ABSTRACT
A 2-y carcinogenicity study of Aloe, Aloe arborescens Miller var. natalensis Berger, a food additive, was conducted for assessment of toxicity and carcinogenic potential in the diet at doses of 4% or 0.8% in groups of male and female Wistar Hannover rats. Both sexes receiving 4% showed diarrhea, with loss of body weight gain. The survival rate in the 4% female group was significantly increased compared with control females after 2 y. Hematological and biochemical examination showed increase of RBC, Hb, and Alb in the 4% males. The cause of these increases could conceivably have been dehydration through diarrhea. AST and Na were significantly decreased in the males receiving 4%, and Cl was significantly decreased in both 4% and 0.8% males. A/G was significantly increased in the 4% females, and Cl was significantly decreased (0.8%) in the female group. Histopathologically, both sexes receiving 4% showed severe sinus dilatation of ileocecal lymph nodes, and yellowish pigmentation of ileocecal lymph nodes and renal tubules. Adenomas or adenocarcinomas in the cecum, colon, and rectum were observed in 4% males but not in the 0.8% and control male groups. Similarly, in females, adenomas in the colon were also observed in the 4% but not 0.8% and control groups. In conclusion, Aloe, used as a food additive, exerted equivocal carcinogenic potential at 4% high-dose level on colon in the 2-y carcinogenicity study in rats. Aloe is not carcinogenic at nontoxic-dose levels and that carcinogenic potential in at 4% high-dose level on colon is probably due to irritation of the intestinal tract by diarrhea.
Sigma-Aldrich
Aloenin, ≥95% (LC/MS-ELSD)