  • Search Results
  • [Clinical, biological and therapeutic profile of anemia by vitamin B12 deficiency in the department of hematology of Marrakech (Morocco)].
[Clinical, biological and therapeutic profile of anemia by vitamin B12 deficiency in the department of hematology of Marrakech (Morocco)].
Bulletin de la Societe de pathologie exotique (1990) (2013-03-29)
H Nafil, I Tazi, M Sifessalam, M Bouchtia, L Mahmal
ABSTRACT
The aim of this study is to examine retrospectively the clinical, biological and treatment features of anemia by vitamin B12 deficiency in the Hematology department of CHU Mohamed VI Marrakech. We report the results of a retrospective study conducted during six years (2005-2010). It included all patients with anemia (with or without thrombocytopenia or leukopenia) associated with vitamin B12 levels <200 pg / ml. One hundred twenty one cases were analyzed. The average age of patients was 62 years (38-89 years) with a female predominance (sex ratio F/M: 1.3). The clinical symptomatology is dominated by pallor (97.5%), cardiovascular signs (46%) and digestive symptoms (34.7%). Neurological signs were noted in 17.3% of cases. The blood count showed anemia (hemoglobin: mean= 6.9 g/dl), macrocythemia (MCV: mean= 109 fl). Leukopenia was noted in 35 patients (29%), thrombocytopenia in 34 patients (28%) and pancytopenia in 21 patients (17,3%). The average vitamin B12 was 72 pg/ml. The causes of B12 deficiency are pernicious anemia (43%), food-cobalamin malabsorption (43%), and in 14% of cases no etiology was found. Gastritis was found in 82.7% of our patients and Helicobacter pylori (HP) infection in 72.7% of cases. Reticulocyte crisis was observed after parenteral administration of hydroxocobalamine within an average of 8 days and normalization of blood counts, in all patients, within an average of 51 days. In patients with HP infection, eradication therapy of HP was performed. The cure rate of the HP is 90%.
Product Description
Hydroxocobalamin, European Pharmacopoeia (EP) Reference Standard
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.