  • Search Results
  • Randomized controlled trial of the effect of depth of anaesthesia on postoperative pain.
Randomized controlled trial of the effect of depth of anaesthesia on postoperative pain.
British journal of anaesthesia (2013-12-11)
C J Law, G M Jacobson, M Kluger, M Chaddock, M Scott, J W Sleigh
ABSTRACT
Our hypothesis was that deep anaesthesia, as estimated by a low target bispectral index (BIS) of 30-40, would result in less postoperative pain than that achieved at a conventional depth of anaesthesia. We undertook a randomized double-blind controlled study at two tertiary teaching hospitals in New Zealand (2010-1) recruiting 135 adult patients ASA I-II presenting for non-emergent surgery under general anaesthesia requiring tracheal intubation. Anaesthesia was maintained with desflurane and a multimodal analgesia regimen comprising fentanyl infusion, i.v. paracetamol, and parecoxib. Patients were randomly assigned to either a low BIS (30-40) group or a high BIS (45-60) group. Desflurane concentrations were titrated to achieve these targets. Postoperative pain was assessed by: the pain on awakening (0-10, verbal rating scale, VRS(awake)) in the post-anaesthetic care unit; pain on activity at 20-24 h after operation (VRS(d1A)); and the rate of morphine patient-controlled analgesia (PCA) usage over the first 24 h. There was no statistically significant difference between the two groups for any of the pain scores. The median [inter-quartile range (IQR)] VRS(awake) was 4.0 (0-8) for the low and 4.0 (0-8) for the high BIS groups (P=0.56). The median (IQR) VRS(d1A) was 3.0 (1-5) for the low and 3.0 (1.5-4.5) for the high BIS groups (P=0.83). The median PCA morphine consumption in the low BIS group was 0.61 mg h(-1) (0.04-1.5) vs 0.43 mg h(-1) (0-1.59) in the high BIS group (P=0.98). We conclude that there is no clinically useful analgesic effect of a deep anaesthesia regimen.
Product Description
Supelco
Fentanyl solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Morphine sulfate salt pentahydrate
Sigma-Aldrich
Fentanyl citrate salt
Supelco
Fentanyl solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine sulfate salt solution, 1.0 mg/mL in methanol, drug standard
Isoflurane, European Pharmacopoeia (EP) Reference Standard