  • Search Results
  • Monitoring protein conformation along the pathway of chaperonin-assisted folding.
Monitoring protein conformation along the pathway of chaperonin-assisted folding.
Cell (2008-04-09)
Shruti Sharma, Kausik Chakraborty, Barbara K Müller, Nagore Astola, Yun-Chi Tang, Don C Lamb, Manajit Hayer-Hartl, F Ulrich Hartl
ABSTRACT
The GroEL/GroES chaperonin system mediates protein folding in the bacterial cytosol. Newly synthesized proteins reach GroEL via transfer from upstream chaperones such as DnaK/DnaJ (Hsp70). Here we employed single molecule and ensemble FRET to monitor the conformational transitions of a model substrate as it proceeds along this chaperone pathway. We find that DnaK/DnaJ stabilizes the protein in collapsed states that fold exceedingly slowly. Transfer to GroEL results in unfolding, with a fraction of molecules reaching locally highly expanded conformations. ATP-induced domain movements in GroEL cause transient further unfolding and rapid mobilization of protein segments with moderate hydrophobicity, allowing partial compaction on the GroEL surface. The more hydrophobic regions are released upon subsequent protein encapsulation in the central GroEL cavity by GroES, completing compaction and allowing rapid folding. Segmental chain release and compaction may be important in avoiding misfolding by proteins that fail to fold efficiently through spontaneous hydrophobic collapse.
Sigma-Aldrich
Atto 647N NHS ester, BioReagent, suitable for fluorescence, ≥85% (coupling to amines)
Sigma-Aldrich
Atto 532 NHS ester, BioReagent, suitable for fluorescence, ≥80% (coupling to amines)
Supelco
Atto 647N, BioReagent, suitable for fluorescence, ≥98.0% (HPCE)
Supelco
Atto 647N maleimide, BioReagent, suitable for fluorescence, ≥90% (coupling to thiols)
Sigma-Aldrich
Atto 532, BioReagent, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Atto 647N-Biotin, BioReagent, ≥95% (HPCE), suitable for fluorescence
Sigma-Aldrich
Atto 647N azide
Sigma-Aldrich
Atto 532 maleimide, BioReagent, suitable for fluorescence, ≥90% (coupling to thiols)
Sigma-Aldrich
Atto 647N iodoacetamide
Sigma-Aldrich
Atto 647N amine
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.