MilliporeSigma
  • Autophagic-lysosomal pathway is the main proteolytic system modified in the skeletal muscle of esophageal cancer patients.
Autophagic-lysosomal pathway is the main proteolytic system modified in the skeletal muscle of esophageal cancer patients.
The American journal of clinical nutrition (2013-10-11)
Nicolas Tardif, Maria Klaude, Lars Lundell, Anders Thorell, Olav Rooyackers
ABSTRACT
In cancer cachexia, muscle depletion is related to morbidity and mortality. Muscle-wasting mechanisms in cancer patients are not fully understood. We investigated the involvement of the proteolytic systems (proteasome, autophagic-lysosomal, calpain, and caspase) in muscle wasting during cancer cachexia. Esophageal cancer patients [n = 14; mean ± SD age: 64.1 ± 6.6 y] and weight-stable control patients undergoing reflux surgery (n = 8; age: 57.5 ± 5.8 y) were included. Enzymatic activities were measured in the vastus lateralis and diaphragm. Protein expressions were also measured in the vastus lateralis of control (n = 7) and cancer (n = 8) patients. Proteasome, calpain, and caspase 3 activities in the vastus lateralis and diaphragm muscles did not differ between the 2 groups. Cathepsin B and L activities were 90% (± SD) [2.4 ± 0.2 compared with 1.3 ± 0.2 pmol 7-amido-4-methylcoumarin (AMC) · μg protein⁻¹ · min⁻¹; P < 0.001] and 115% (5.3 ± 0.4 compared with 2.5 ± 0.3 pmol AMC · μg protein⁻¹ · min⁻¹; P < 0.001) greater, respectively, in the vastus lateralis of cancer patients than in that of control subjects. We observed (in conjunction with increased lysosomal protease activities) higher microtubule-associated protein 1 light chain 3B-II/I ratios (0.14 ± 0.08 compared with 0.04 ± 0.04) and cathepsin B and L expressions in the vastus lateralis of cancer patients than in that of control subjects (P < 0.05). Protein expression of p62 in the vastus lateralis did not differ between the 2 groups. The autophagic-lysosomal pathway in the skeletal muscle of cancer patients was modified, whereas other proteolytic systems were unchanged. These findings suggest involvement of the autophagic-lysosomal proteolytic system during cancer cachexia development in humans.
Sigma-Aldrich
Cathepsin B from bovine spleen, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
Monoclonal Anti-Cathepsin L antibody produced in mouse, clone 33/2, ascites fluid
Sigma-Aldrich
Cathepsin B from human liver, buffered aqueous solution, ≥1,500 units/mg protein (E1%/280)
Sigma-Aldrich
Anti-Cathepsin B (Ab-1) Mouse mAb (CA10), lyophilized, clone CA10, Calbiochem®
Sigma-Aldrich
Cathepsin L from human liver, ≥0.5 units/mg protein, solution
Sigma-Aldrich
Cathepsin B from human placenta, lyophilized powder, ≥5 units/mg protein