  • Search Results
  • Endothelial cellular senescence is inhibited by liver X receptor activation with an additional mechanism for its atheroprotection in diabetes.
Endothelial cellular senescence is inhibited by liver X receptor activation with an additional mechanism for its atheroprotection in diabetes.
Proceedings of the National Academy of Sciences of the United States of America (2014-01-09)
Toshio Hayashi, Hitoshi Kotani, Tomoe Yamaguchi, Kumiko Taguchi, Mayu Iida, Koichiro Ina, Morihiko Maeda, Masafumi Kuzuya, Yuichi Hattori, Louis J Ignarro
ABSTRACT
Senescence of vascular endothelial cells leads to endothelial dysfunction and contributes to the progression of atherosclerosis. Liver X receptors (LXRs) are nuclear receptors whose activation protects against atherosclerosis by transcriptional regulation of genes important in promoting cholesterol efflux and inhibiting inflammation. Here we found that LXR activation with specific ligands reduced the increase in senescence-associated (SA) β-gal activity, a senescence marker, and reversed the decrease in telomerase activity, a replicative senescence marker, in human endothelial cells under high glucose. This effect of LXR activation was associated with reduced reactive oxygen species and increased endothelial NO synthase activity. A series of experiments that used siRNAs indicated that LXRβ mediates the prevention of endothelial cellular senescence, and that sterol regulatory element binding protein-1, which was up-regulated as a direct LXRβ target gene, may act as a brake of endothelial cellular senescence. Although oral administration of the LXR ligand led to severe fatty liver in diabetic rats, concomitant therapy with metformin avoided the development of hepatic steatosis. However, the preventive effect of the LXR ligand on SA β-gal-stained cells in diabetic aortic endothelium was preserved even if metformin was coadministered. Taken together, our studies demonstrate that an additional mechanism, such as the regulation of endothelial cellular senescence, is related to the antiatherogenic properties of LXRs, and concomitant treatment with metformin may provide a clinically useful therapeutic strategy to alleviate an LXR activation-mediated adverse effects on liver triglyceride metabolism.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.