  • Search Results
  • Noncanonical functions of telomerase: implications in telomerase-targeted cancer therapies.
Noncanonical functions of telomerase: implications in telomerase-targeted cancer therapies.
Cancer research (2014-03-07)
Yinghui Li, Vinay Tergaonkar
ABSTRACT
Telomerase plays a pivotal role in bypassing cellular senescence and maintaining telomere homeostasis, essential properties required for the sustenance and progression of cancer. However, recent investigations have uncovered extratelomeric properties of telomerase that are independent of its role in telomere extension. This review summarizes recent insights to the noncanonical functions of telomerase reverse transcriptase (TERT) catalytic subunit, in particular in cancer progression, and highlights two major signaling mechanisms involved in the cross-talk with TERT-the NF-κB and Wnt/β-catenin pathways. We propose a feed-forward regulatory loop mechanism underlying TERT activation in cancers in which TERT acts as a transcriptional modulator of oncogenic signaling pathways that sustain its own levels and control the induction of target genes critical for tumor cell survival and proliferation. Finally, we provide a new perspective on telomerase-targeted cancer therapies and suggest possible interventions targeting the nontelomeric roles of TERT. This therapeutic strategy can be used in the future targeting of other telomerase components that exhibit novel nontelomeric functions in cancer and other ailments.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.