  • Search Results
  • Crystal structure analysis of EstA from Arthrobacter sp. Rue61a--an insight into catalytic promiscuity.
Crystal structure analysis of EstA from Arthrobacter sp. Rue61a--an insight into catalytic promiscuity.
FEBS letters (2014-03-13)
Ulrike Gabriella Wagner, Frank DiMaio, Stephan Kolkenbrock, Susanne Fetzner
ABSTRACT
In this article we analyze the reasons for catalytic promiscuity of a type VIII esterase with β-lactamase fold and the ability to cleave β-lactams. We compared the structure of this enzyme to those of an esterase of the same type without any lactamase ability, an esterase with moderate lactamase ability, and a class C β-lactamase with similar fold. Our results show that for these enzymes, the difference in the substrate specificity is sterically driven.
Sigma-Aldrich
β-Lactamase, recombinant, expressed in E. coli
Sigma-Aldrich
β-Lactamase from Pseudomonas fluorescens, lyophilized powder, recombinant, expressed in E. coli
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized powder, 1,500-3,000 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
β-Lactamase from Enterobacter cloacae, Type IV, lyophilized powder, 0.2-0.6 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
β-Lactamase from Enterobacter cloacae, Type III, lyophilized powder, 6-18 units/mg protein (using benzylpenicillin)
Sigma-Aldrich
Penicillinase from Bacillus cereus, lyophilized, powder, white, ~13 U/mg
Sigma-Aldrich
β-Lactamase Blend, recombinant, expressed in E. coli