  • Search Results
  • [Cloning and analysis of reverse transcriptase(RT) of Ty1-copia retrotransposons in Dendrobium officinale].
[Cloning and analysis of reverse transcriptase(RT) of Ty1-copia retrotransposons in Dendrobium officinale].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2014-04-26)
Cong Li, Jin-Ping Si, Yan-Hui Gao, Yu-Qiu Zhu
ABSTRACT
Using universal primer Ty1-copia retrotransposon RT,43 Ty1-copia like retrotransposon RT with high heterogeneity, stop codon mutation and frameshift mutation were amplified by PCR from genomic DNA of Zhejiang Lin'an (C15) and Yunnan Guangnan (A39) of Dendrobium officinale. The length of these sequences varied from 260 to 266 bp, and was rich in AT and consistency ranged from 47.1% to 97.7%. Different c/s-acting regulatory elements induced by low temperature, heat, light, all kinds of plant growth regulating substances and the starting transcription signals, corresponding to CAAT box, TATA box conserved sequences and some other regulatory elements. When being translated into amino acids, ten sequences presented stop codon mutation, five sequences presented frameshift mutation, and thirty-seven sequences presented conserved sequence "SLYGKQ" mutation. Six categories were identified through phylogenic analysis after alignment analyses of their amino acid sequences, and with other plants (eg. Triticum aestivum, Eleocharis quinqueflora) having high homology, which indicated that horizontal transmission of retrotransposon occurred among the plants in the past.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.